DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon sperm)

Synonyms: DNA sodium saltThymonucleic acid sodium salt

J14377-22 J14377-A1
101175-480EA 123.07 USD
101175-480 101175-478
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon sperm)
DNA sodium salt (Deoxyribonucleic acid sodium salt)
Storage Temperature: Ambient
MDL Number: MFCD00284534
CAS Number: 9007-49-2
Merck Index: 14,02907

Order Now

Specification Test Results

Assay (unspecified), Protein ≤5.0%
Impurity Content, Phosphorous ≤9.0%
Loss on Drying ≤18.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR