Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) dihydrate ≥96.0% (by UV/Vis)
Supplier: Thermo Fisher Scientific
Synonyms:
Cytidine-5'-Triphosphate disodium salt dihydrate
, Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate dihydrate
J14121-ME
101174-324EA
325.26
USD
101174-324
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) dihydrate ≥96.0% (by UV/Vis)
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) dihydrate
Specification Test Results
Assay (UV/vis) | ≥97.5% |
Epsilonmax (pH 7, NRC) | 9.1×10³ |
Epsilon₂₆₀ (pH 7, NRC) | 7.55×10³ |
Lambdamax (pH 7, NRC) | 271 nm |
Identification (FTIR) | Conforms |
Loss on Drying | ≤1.0% |
lambda-max (UV-Vis), pH 1.0 | 280 ±2 nm |
Spectral Ratio, A₂₅₀/A₂₆₀ (pH 2.0) | 0.45 ±0.02 |
Spectral Ratio, A₂₈₀/A₂₆₀ (pH 2.0) | 2.10 ±0.05 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...