4-Aminoantipyrine 98+%

Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one4-Aminophenazone

J11056-18 J11056-22
101174-198EA 95.49 USD
101174-198 101174-196
4-Aminoantipyrine 98+%
4-Aminoantipyrine
Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 14,00591

Order Now

Specification Test Results

Assay ≥98.0%
Identification (FTIR) Conforms
Solution Test (5%, H₂O) Clear Solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR