Agar

Supplier: Thermo Fisher Scientific

Synonyms: Agar Agar

J10654-Q1 J10654-P2 J10654-P5
101169-724EA 1973.91 USD
101169-724 101174-168 101172-128
Agar
Agar
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,00184
https://us.vwr.com/stibo/web/std.lang.all/81/91/2298191.jpg

Order Now

Specification Test Results

Total Ash ≤6.5%
Loss on Drying ≤20.0%
Identity Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR