Venturi V26 LX Series General Purpose Bench Fume Hoods with 28" Combination Vertical/Horizontal Sash, Phenolic Resin Liner, Kewaunee Scientific
Supplier: Kewaunee
V26F283060TM V26F283072TS V26F282472TS V26F283096TM V26F283096TS V26F283696TS V26F283060TS V26F282448TM V26F283048TS V26F283660TS V26F283648TS V26F283048TM V26F283648TM V26F282460TM V26F282472TM V26F283672TM V26F283072TM V26F282496TS V26F283672TS V26F282460TS V26F282496TM V26F283660TM V26F283696TM V26F282448TS
KSV26F283672TMEA 24457.15 USD
KSV26F283672TM KSV26F283660TM KSV26F283660TS KSV26F283672TS KSV26F283048TS KSV26F283696TM KSV26F283648TM KSV26F283048TM KSV26F283096TS KSV26F283696TS KSV26F283060TS KSV26F282460TM KSV26F283072TS KSV26F282496TM KSV26F283096TM KSV26F283648TS KSV26F282472TM KSV26F283060TM KSV26F282472TS KSV26F282460TS KSV26F283072TM KSV26F282448TM KSV26F282496TS KSV26F282448TS
Venturi V26 LX Series General Purpose Bench Fume Hoods with 28" Combination Vertical/Horizontal Sash, Phenolic Resin Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Ordering information: Work top and base cabinets not included with fume hood.