Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, Phenolic Resin Liner, Kewaunee Scientific
Supplier: Kewaunee
V25F283696TM V25F283072TM V25F283048TM V25F283072TN V25F283672TS V25F282472TS V25F282448TN V25F283672TN V25F283648TN V25F283096TS V25F282472TN V25F282460TS V25F283660TS V25F282460TN V25F283648TM V25F283060TN V25F283648TS V25F283096TM V25F283672TM V25F283696TS V25F283660TN V25F282496TM V25F283048TN V25F282496TN V25F283660TM V25F283696TN V25F283060TM V25F283096TN V25F283060TS V25F282460TM V25F282448TS V25F282448TM V25F283072TS V25F282496TS V25F283048TS V25F282472TM
KSV25F282496TSEA 27078.8 USD
KSV25F282496TS KSV25F283048TM KSV25F283048TN KSV25F283060TS KSV25F282472TM KSV25F283648TS KSV25F282496TM KSV25F282472TN KSV25F282496TN KSV25F282472TS KSV25F283048TS KSV25F283696TS KSV25F283672TS KSV25F282448TS KSV25F283060TN KSV25F283672TM KSV25F283672TN KSV25F283696TM KSV25F283696TN KSV25F283060TM KSV25F283096TS KSV25F283072TS KSV25F282460TM KSV25F282460TN KSV25F282448TN KSV25F282448TM KSV25F282460TS KSV25F283648TM KSV25F283648TN KSV25F283072TN KSV25F283096TM KSV25F283660TM KSV25F283072TM KSV25F283096TN KSV25F283660TN KSV25F283660TS
Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, Phenolic Resin Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Ordering information: Work top and base cabinets not included with fume hood.