Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, KMER Liner, Kewaunee Scientific
Supplier: Kewaunee
V25F282460KN V25F282460KM V25F283096KN V25F282496KN V25F283060KM V25F283060KN V25F283048KN V25F283648KN V25F282496KM V25F283696KS V25F283660KM V25F283660KN V25F282448KN V25F283696KM V25F283648KS V25F282472KS V25F282448KM V25F283072KS V25F282460KS V25F283696KN V25F283096KS V25F283096KM V25F282472KM V25F283672KN V25F283072KM V25F282448KS V25F283660KS V25F283648KM V25F282472KN V25F283048KM V25F283060KS V25F283672KM V25F283048KS V25F283072KN V25F282496KS V25F283672KS
KSV25F283072KMEA 20799.75 USD
KSV25F283072KM KSV25F282460KM KSV25F283096KM KSV25F282460KN KSV25F283072KN KSV25F282460KS KSV25F283696KS KSV25F283096KS KSV25F283672KS KSV25F283096KN KSV25F283072KS KSV25F283048KS KSV25F283660KN KSV25F283660KM KSV25F282472KS KSV25F282496KS KSV25F283060KM KSV25F283060KN KSV25F283648KS KSV25F282472KM KSV25F282472KN KSV25F282496KN KSV25F282496KM KSV25F283660KS KSV25F283048KM KSV25F283048KN KSV25F283648KM KSV25F283648KN KSV25F283060KS KSV25F283672KM KSV25F282448KN KSV25F282448KM KSV25F283696KN KSV25F283672KN KSV25F283696KM KSV25F282448KS
Venturi V25 LX Series General Purpose Bench Fume Hoods with 28" Vertical Rising Sash, KMER Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Ordering information: Work top and base cabinets not included with fume hood.