Venturi V06 General Purpose Bench Fume Hood with Combination Vertical Rising/Horizontal Sash, Phenolic Resin Liner, Kewaunee Scientific
Supplier: Kewaunee
V06F283060TM V06F282496TS V06F283060TS V06F283660TS V06F283648TM V06F282448TM V06F282460TM V06F283660TM V06F283096TS V06F283672TM V06F283696TM V06F282472TS V06F283696TS V06F282472TM V06F283672TS V06F283072TS V06F283648TS V06F282460TS V06F282448TS V06F283048TS V06F283072TM V06F282496TM V06F283096TM V06F283048TM
KSV06F282448TMEA 14958.35 USD
KSV06F282448TM KSV06F283672TM KSV06F283048TM KSV06F283660TM KSV06F283096TS KSV06F282448TS KSV06F282496TM KSV06F282460TS KSV06F282496TS KSV06F283048TS KSV06F282472TS KSV06F283648TM KSV06F282460TM KSV06F283648TS KSV06F282472TM KSV06F283696TM KSV06F283072TS KSV06F283060TS KSV06F283696TS KSV06F283072TM KSV06F283060TM KSV06F283660TS KSV06F283096TM KSV06F283672TS
Venturi V06 General Purpose Bench Fume Hood with Combination Vertical Rising/Horizontal Sash, Phenolic Resin Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Ordering information: Work top and base cabinets not included with fume hood.