Venturi V05-General Purpose Bench Fume Hood with Vertical Rising Sash, Type 304L Stainless Steel Liner, Kewaunee Scientific
Supplier: Kewaunee
V05F282460SS V05F283648SN V05F283660SN V05F282448SN V05F282472SS V05F282448SM V05F283696SN V05F282496SN V05F282496SM V05F282448SS V05F283660SM V05F283696SM V05F283060SS V05F283696SS V05F283072SN V05F283096SS V05F283648SS V05F283048SS V05F282460SN V05F283060SM V05F283672SS V05F283096SN V05F283060SN V05F283648SM V05F283672SN V05F282472SN V05F283096SM V05F283048SM V05F283672SM V05F283072SM V05F283048SN V05F283072SS V05F282472SM V05F283660SS V05F282496SS V05F282460SM
KSV05F283072SNEA 17329.4 USD
KSV05F283072SN KSV05F283096SM KSV05F282460SM KSV05F283096SN KSV05F283072SM KSV05F283660SM KSV05F283660SN KSV05F282448SS KSV05F282448SN KSV05F283660SS KSV05F282448SM KSV05F282460SS KSV05F283072SS KSV05F283096SS KSV05F283648SS KSV05F282460SN KSV05F283060SN KSV05F283060SM KSV05F282496SM KSV05F282496SN KSV05F283648SN KSV05F283648SM KSV05F282496SS KSV05F283048SS KSV05F283672SM KSV05F283696SM KSV05F283672SN KSV05F283696SN KSV05F283672SS KSV05F283696SS KSV05F282472SS KSV05F283060SS KSV05F282472SN KSV05F283048SM KSV05F283048SN KSV05F282472SM
Venturi V05-General Purpose Bench Fume Hood with Vertical Rising Sash, Type 304L Stainless Steel Liner, Kewaunee Scientific
Fume Hoods and Enclosures Chemical Fume Hoods
The Supreme Air Venturi provides the safest user-friendly fume hood manufactured today.
  • Un paralleled Venturi enhanced containment technology
  • High performance/energy efficient
  • Unique ultra-smooth sash system
  • Exceptional viewing height
  • Flush airfoil
  • LED lights with color and intensity adjustability
  • Electromechanical sash stops
The Supreme Air Venturi utilizes proven containment enhancing techniques coupled with innovative laboratory convenience features, including: adjustable LED lights, electromechanical sash stops, ultra-smooth sash system, and push button sash management.
Certifications: UL and cUL listed. Tested to ASHRAE 110, NIH03-112C, ASTM D552, and NFPA 45.
Ordering information: Work top and base cabinets not included with fume hood.