PIPETMAN® M Multichannel Electronic Pipettor, Gilson®
F81026 F81030 F81020 F81028 F81025 F81029 F81032 F81021 F81018 F81019 F81031 F81027
76178-398EA 1743.98 USD
76178-398 76178-400 76178-090 76178-402 76178-092 76178-404 76178-094 76178-096 76178-084 76178-086 76178-088 76178-396
PIPETMAN® M Multichannel Electronic Pipettor, Gilson®
Motorized air-displacement multichannel pipettor PIPETMAN® M.
  • Electronic pipettor
  • Guaranteed performance
  • Enhanced productivity and comfort with guaranteed performance in standard and repetitive modes
PIPETMAN® M Multichannel Pipettors boast enhanced ergonomic features and new motorized user-friendly functionality for higher throughput. The signature Gilson Trident is featured to create a stepped ejection, to significantly reduce the force required to eject tips. Lightweight and extremely comfortable, the PIPETMAN® M Multichannel Pipettor is a fully motorized device that requires virtually zero pipetting forces to aspirate and dispense samples. Simply turn the push-button to adjust settings and then click. Four essential pipetting modes are included; reverse, mix, repetitive, and pipetting.
Certifications: Gilson volumetric specifications in standard pipetting (PIPET mode) are guaranteed with a performance exceeding ISO 8655-2 recommendations for this mode. In the absence of ISO recommendations for repetitive pipetting mode for air-displacement pipettes, Gilson volumetric specifications for repetitive pipetting (REPETITIVE mode) are guaranteed within ISO 8655-2 recommendations for standard pipetting (Cf. ISO 8655-2 table 1).