Vortec Pro®, Thin Grinding Wheel, 27, Weiler
Supplier: Weiler
Vortec Pro®
804-56389
76184-172EA 3.99 USD
76184-172
Vortec Pro®, Thin Grinding Wheel, 27, Weiler
Abrasives
Resin-bonded disc is formulated for aggressive stock removal and fast cutting
  • Reinforced with fiberglass webbing for stability
Each pack includes twelve heads and handles.
Suitable for general purpose cutting and grinding applications on steel, iron, and other ferrous metals.