trans,trans-2,4-Decadienal ≥95%

Synonyms: Decadienal(2E,4E)-Deca-2,4-dienal

BML-HA005-0020
89160-050EA 90.02 USD
89160-050
trans,trans-2,4-Decadienal ≥95%
trans,trans-2,4-Decadienal

Induces mitochondrial dysfunction and oxidative stress.
Decadienal is a lipid peroxidation product which is an aldehydic bifuntional electrophile. It induces mitochondrial dysfunction and oxidative stress possibly via covalent modification of cytochrome c. It can induce cell proliferation via the p27 pathway in human bronchial epithelial cells.

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
MDL Number: MFCD00007007
CAS Number: 25152-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR