Phenylbutazone ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 4-Butyl-1,2-diphenylpyrazolidine-3,5-dione

430-112-G005
89151-126EA 42.47 USD
89151-126
Phenylbutazone ≥98% (by HPLC)
Phenylbutazone

Anti-inflammatory agent
Anti-inflammatory agent. Substrate for prostaglandin peroxidase.

Formula: C₁₉H₂₀N₂O₂
MW: 308.38 g/mol
MDL Number: MFCD00005500
CAS Number: 50-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR