Galvinoxyl ≥95% (by TLC) free radical

400-016-M050 400-016-M010
89150-592EA 44.39 USD
89150-592 89150-590
Galvinoxyl ≥95% (by TLC) free radical
Galvinoxyl

Free radical compound.

Formula: C₂₉H₄₁O₂
MDL Number: MFCD00001616
CAS Number: 2370-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR