UltraBrite™ Black IHC chromogen (HRP), BioVision
Supplier: BioVision
M1311-30
76181-788EA 437.81 USD
76181-788
UltraBrite™ Black IHC chromogen (HRP), BioVision
Unique black IHC chromogen for HRP-activated development.
  • APPLICATIONS: IHC, ISH (in situ hybridization)
UltraBrite™ black IHC chromogen (HRP) is a substrate-chromogen system designed to be used for either immunohistochemistry (IHC) or ISH when utilizing horseradish peroxidase. UltraBrite™ black IHC chromogen (HRP) produces a sharply contrasting ebony black color that can easily be distinguished from other stains.