Anti-CD3E Goat Polyclonal Antibody

Supplier: BioVision
A1386-100
76177-970EA 344.42 USD
76177-970
Anti-CD3E Goat Polyclonal Antibody
The CD3 complex mediates signal transduction, resulting in T-cell activation and proliferation. Required for normal immune responses.

Western blot: 0.1 µg/ml

Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR