Would you like to log in again?
VWR® Sterile PETG Single-Use Bottle with Aseptic Connector
Supplier: VWR International
VWR®
5000899 5000898 5000895 5000897 5000896
Sterile PETG containers provide the maximum amount of flexibility and reliability.
  • Platinum cured silicone tubing
  • CPC® AseptiQuik® S Connector
  • USP Class VI
  • Animal-free or processed in accordance to EMEA/410/01
  • Assemblies are manufactured in certified ISO Class 7 cleanroom
  • Gamma irradiated to Sterility Assurance Leve (SAL) 10⁻⁶
  • Each assembly comes individually double-bagged
  • Multiple bottle sizes allow for process-specific flexibility
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and applications. Using assemblies with an aseptic connector eliminates the need for a laminar hood as well as additional equipment (ie. welder), lessens the risk of contamination, and saves valuable time. The AseptiQuik® “Flip-Click-Pull” design enables users to quickly and easily transfer small volumes of media. When used with sterile aseptic connector tube sets, custom possibilities are endless. Other aseptic connectors are available upon request. Please consult a VWR® Account Manager to discuss custom single-use needs.
ORDER
Expand All / Collapse All
Description Material Capacity Cap Size VWR Catalog Number Unit Price Quantity
Port 1 Tubing Connection: AQS; Port 2 Tubing Connection: 25 mm Hydrophobic PE Vent Filter PETG Bottle, Silicone Tubing 125 mL (4 oz.) 38-430 76169-206 Case of 24 Retrieving
Pack of 6 Retrieving
Port 1 Tubing Connection: AQS; Port 2 Tubing Connection: 25 mm Hydrophobic PE Vent Filter PETG Bottle, Silicone Tubing 250 mL (8 oz.) 38-431 76169-208 Case of 24 Retrieving
Pack of 6 Retrieving
Port 1 Tubing Connection: AQS; Port 2 Tubing Connection: 25 mm Hydrophobic PE Vent Filter PETG Bottle, Silicone Tubing 500 mL (16 oz.) 38-432 76169-210 Case of 24 Retrieving
Pack of 6 Retrieving
Port 1 Tubing Connection: AQS; Port 2 Tubing Connection: 25 mm Hydrophobic PE Vent Filter PETG Bottle, Silicone Tubing 1000 mL (33 oz.) 38-433 76169-196 Case of 12 Retrieving
Pack of 3 Retrieving
Port 1 Tubing Connection: AQS; Port 2 Tubing Connection: 50 mm Hydrophobic PE Vent Filter PETG Bottle, Silicone Tubing 2000 mL (67 oz.) 53B 76169-198 Case of 12 Retrieving
Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Tubing size Port 1: 1/8 x 1/4"; Port 2: 1/8 x 3/8" Port 1: 1/8 x 1/4"; Port 2: 1/8 x 1/4"
Tubing length Port 1: 12" Port 2: 3"
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty