L-655,240 ≥98% (by HPLC)
BML-RA117-0005
89162-522EA
333.45
USD
89162-522
L-655,240 ≥98% (by HPLC)
L-655,240
Thromboxane receptor ligand
Potent, selective, orally active thromboxane A2 (TXA2) receptor antagonist (IC50=7nM).
Formula:
C₂₁H₂₁ClFNO₂ MW: 373.9 g/mol |
CAS Number:
103253-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...