JX-401 ≥98% (by TLC)
BML-EI366-0001
BML-EI366-0010
89158-196EA
205.38
USD
89158-196
89158-194
JX-401 ≥98% (by TLC)
JX-401
p38 inhibitor
Benzylpiperidine inhibitor of p38α (IC50 = 32 nM). Selective over p38γ (IC50 10 µM).
Formula:
C₂₁H₂₅NO₂S MW: 355.5 g/mol |
CAS Number:
349087-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...