APHS ≥98% (by TLC)

BML-EI339-0050 BML-EI339-0010
89158-116EA 462.76 USD
89158-116 89158-114
APHS ≥98% (by TLC)
APHS

COX inhibitor


A selective COX-2 inhibitor (IC50=0.8 µM) that covalently modifies the enzyme by acetylation of Ser530. APHS is an inhibitor of HIV-1 replication.

Formula: C₁₅H₁₈O₂S
MW: 262.4 g/mol
CAS Number: 209125-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR