ASSAD111
BML-D111-0010
89157-308EA 112.01 USD
89157-308
ASSAD111
Dopamine receptor ligand
A potent and selective dopamine D2 antagonist. In animals it displays minimal cataleptic effects at doses that block dopamine agonist-induced hyperactivity. An atypical antipsychotic agent.