AM 1241 ≥97% (by TLC)

550-383-M001 550-383-M005
89152-652EA 173.7 USD
89152-652 89152-650
AM 1241 ≥97% (by TLC)
AM 1241

CB2 receptor agonist


Potent and selective CB2 receptor agonist (Ki=3.4nM (mouse), Ki=280nM (rat) also in vivo.

Formula: C₂₂H₂₂N₃O₃I
MW: 503.3 g/mol
CAS Number: 444912-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR