JYL-79
Synonyms:
Vanilloid Receptor Agonist 23
, 2-(3,4-Dimethylbenzyl)-3-{[(4-hydroxy-3-methoxy-benzyl)amino]carbothioyl}propyl pivalate
550-296-M001
89152-484EA
66.9
USD
89152-484
JYL-79
JYL-79
TRPV1 agonist
Potent TRPV1 agonist (Ki=19nM). About 300-fold more potent than capsaicin (Prod. No. BML-EI125). RTX binding affinity (Ki=17.4nM), agonism (calcium influx; EC50=1.97nM).
Formula:
C₂₆H₃₆N₂O₄S |
CAS Number:
289902-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...