(S)-3C4H-PG ≥97%

550-197-M001
89152-316EA 51.98 USD
89152-316
(S)-3C4H-PG ≥97%
(S)-3C4H-PG

Mixed mGluR agonist and antagonist


Mixed group I mGluR antagonist and group II mGluR agonist.

Formula: C₉H₉NO₅·H₂O
MW: 229.2 g/mol
CAS Number: 55136-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR