N-Benzylnaltrindole hydrochloride ≥95%

550-164-M001 550-164-M005
89152-262EA 104.93 USD
89152-262 89152-260
N-Benzylnaltrindole hydrochloride ≥95%
N-Benzylnaltrindole hydrochloride

δ2-opioid receptor antagonist


Potent δ2-selective opioid receptor antagonist with a long duration of action in vivo.

Formula: C₃₃H₃₂N₂O₃·HCl
MW: 504.6 · 36.5 g/mol
CAS Number: 1206487-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR