(S)-4C3H-PG ≥98% (by HPLC)

550-119-M001
89152-194EA 44.39 USD
89152-194
(S)-4C3H-PG ≥98% (by HPLC)
(S)-4C3H-PG

Mixed mGluR agonist and antagonist


Agonist for group II mGluRs. Competitive antagonist for group I mGluR1; mixed effects at mGluR5.

Formula: C₉H₉NO₅
MW: 211.2 g/mol
CAS Number: 85148-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR