R5573 hydrochloride ≥99%

550-107-M005 550-107-M001 550-107-M025
89152-168EA 566.74 USD
89152-168 89152-166 89152-164
R5573 hydrochloride ≥99%
R5573 hydrochloride

Spirodecanone-site blocker.


Off-white powder.


Soluble in DMSO, dimethyl formamide or methanol.

Formula: C₂₈H₃₁N₃O·HCl
MW: 425.6 · 36.5 g/mol
Storage Temperature: Refrigerator
CAS Number: 1060-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR