MCPIO ≥98% (by HPLC)
430-083-M010
89151-050EA 15.22 USD
89151-050
MCPIO ≥98% (by HPLC)
MCPIO
Spin trapping reagent
  • Soluble in water, 100% ethanol or DMSO.
Cell permeable, stable spin trap. Alternative to PBN (Prod. No. ALX-430-082).