Allethrin ≥95% (by HPLC)

430-031-M025 430-031-M010
89150-932EA 59.6 USD
89150-932 89150-930
Allethrin ≥95% (by HPLC)
Allethrin

Type I pyrethroid, useful as a very weakly active "negative" control for inhibition of calcineurin (protein phosphatase 2B) by type II pyrethroids.

Formula: C₁₉H₂₆O₃
MW: 302.4 g/mol
CAS Number: 584-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR