Aminoguanidinium hydrogen carbonate ≥97%

Synonyms: 1-Aminoguanidinium hydrogen carbonateAminoguanidine hydrogen carbonate

420-004-M500
89150-696EA 44.39 USD
89150-696
Aminoguanidinium hydrogen carbonate ≥97%
Aminoguanidinium hydrogen carbonate

NOS inhibitor


Inhibitor of nitric oxide synthase (NOS).

Formula: C₂H₈N₄O₃
MW: 136.11 g/mol
MDL Number: MFCD00012949
CAS Number: 2582-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR