Encecalin ≥96% (by HPLC)

350-393-M001
89149-682EA 195.25 USD
89149-682
Encecalin ≥96% (by HPLC)
Encecalin

Antifungal agent


Isolated from Encelia farinosa.


Exhibits antifungal activity. Putative compound for the treatment of the dermatophytes Trichophyton rubrum and T. mentagrophytes, the causative agents of tinea pedis infection. Has also been shown to exhibit phytotoxic effects by inhibiting the photosystem I (but not photosystem II) of isolated spinach chloroplasts in a dose dependent manner. The site of inhibition has been located in the span from P680 to QA of the electron transport chain.

Formula: C₁₄H₁₆O₃
CAS Number: 20628-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR