2-Bromoaldisine ≥96% (by HPLC)

350-388-MC05
89149-668EA 149.62 USD
89149-668
2-Bromoaldisine ≥96% (by HPLC)
2-Bromoaldisine

Inhibitor of the Raf/MEK-1/MAPK cascade


Isolated from sponge Pseudaxynissa sp.


Inhibitor of the Raf/MEK-1/MAPK cascade (IC50= 539nM).

Formula: C₈H₇BrN₂O₂
MW: 243.1 g/mol
CAS Number: 96562-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR