Angoroside C ≥98% (by HPLC)

350-331-M001 350-331-M005
89149-514EA 44.39 USD
89149-514 89149-516
Angoroside C ≥98% (by HPLC)
Angoroside C

Antioxidant


Isolated from the root of Scrophularia ningpoensis Hemsl (Figwort).


Potential anti-inflammatory compound. Inhibitor of prostaglandin E2 release in mouse peritoneal macrophages in vitro. Shows potent antioxidative activity in reducing the oxidized OH adducts of dAMP and dGMP. Reveals some trypanocidal potential.

Formula: C₃₆H₄₈O₁₉
MW: 784.8 g/mol
CAS Number: 115909-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR