Carazostatin

350-253-M001 350-253-C100
89149-338EA 72.28 USD
89149-338 89149-340
Carazostatin
Carazostatin

Free radical scavenger


Isolated from Streptomyces chrestomyceticus.


Antioxidant. Free radical scavenger. Potent inhibitor of lipid peroxidation.

Formula: C₂₀H₂₅NO
MW: 295.4 g/mol
CAS Number: 126168-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR