Dammarenolic acid ≥97% (by TLC)

350-155-M001 350-155-M005
89149-190EA 126.78 USD
89149-190 89149-192
Dammarenolic acid ≥97% (by TLC)
Dammarenolic acid

Antiviral agent


Isolated from Aglaia sp.


Dammarane triterpene. Shows antiviral activity with relatively low cytotoxicity.

Formula: C₃₀H₅₀O₃
MW: 458.7 g/mol
CAS Number: 34336-09-9
https://us.vwr.com/stibo/web/std.lang.all/76/91/45347691.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR