Olomoucine, Iso ≥98% (by TLC)

350-090-M005 350-090-M025
89149-032EA 135.67 USD
89149-032 89149-034
Olomoucine, Iso ≥98% (by TLC)
Olomoucine, Iso

Negative control for olomoucine


Inactive negative control for use in studies involving olomoucine (Prod. No. ALX-350-013).

Formula: C₁₅H₁₈N₆O
MW: 298.3 g/mol
CAS Number: 101622-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR