HSD inhibitor 23 ≥95%
270-474-M001
89148-024EA
302.77
USD
89148-024
HSD inhibitor 23 ≥95%
HSD inhibitor 23
11-β-hydroxysteroid dehydrogenase inhibitor
Selective, potent and cell permeable small molecule inhibitor of 11-β-hydroxysteroid dehydrogenase.
Formula:
C₂₄H₃₃F₃N₄O₂ MW: 466.5 g/mol |
CAS Number:
868604-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...