N-(4-Aminobutyl)-5-chloronaphthalene-2-sulfonamide hydrochloride ≥98% (by TLC)

Synonyms: W-13 Hydrochloride

270-176-M005
89147-328EA 119.19 USD
89147-328
N-(4-Aminobutyl)-5-chloronaphthalene-2-sulfonamide hydrochloride ≥98% (by TLC)
N-(4-Aminobutyl)-5-chloronaphthalene-2-sulfonamide hydrochloride

Calmodulin antagonist


Isomer of W-13 (Prod. No. ALX-270-156).

Formula: C₁₄H₁₇ClN₂O₂S·HCl
MW: 349.27 g/mol
MDL Number: MFCD00058030
CAS Number: 88519-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR