S-Isopropyl-ITU hydrobromide ≥98%

270-028-M010 270-028-M050
89147-082EA 19.03 USD
89147-082 89147-084
S-Isopropyl-ITU hydrobromide ≥98%
S-Isopropyl-ITU hydrobromide

NOS inhibitor


Inhibitor of nitric oxide synthase (NOS). 10 times more potent than S-Methyl-ITU (Prod. No. ALX-270-027).

Formula: C₄H₁₀N₂S·HBr
MW: 199.1 g/mol
CAS Number: 4269-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR