S-Methylisothiouronium sulfate ≥97%

Synonyms: 2-Methyl-2-thiopseudourea sulfateS-Methylisothiourea sulfate2-Methyl-2-thiopseudourea hemisulfateS-Methylisothiourea sulphateS-Methylthiouronium sulfateS-Methylthiouronium sulphate2-Methyl-2-thiopseudourea sulphate2-Methyl-2-thiopseudourea hemisulphate

270-027-M250
89147-080EA 44.39 USD
89147-080
S-Methylisothiouronium sulfate ≥97%
S-Methylisothiouronium sulphate

iNOS inhibitor


Reported to be 10- to 30-fold more potent than L-NMMA (Prod. No. ALX-106-001) as an inhibitor of inducible nitric oxide synthase (iNOS; NOS II).

Formula: C₄H₁₂N₄S₂·H₂SO₄
MW: 278.38 g/mol
MDL Number: MFCD00013055
CAS Number: 867-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR