(S)-(-)-2-Guanidinoglutaric acid ≥98%

270-023-M010
89147-064EA 77.35 USD
89147-064
(S)-(-)-2-Guanidinoglutaric acid ≥98%
(S)-(-)-2-Guanidinoglutaric acid

nNOS inhibitor


Linear mixed-type inhibitor of neuronal nitric oxide synthase (nNOS/NOS I).

Formula: C₆H₁₁N₃O₄
MW: 189.17 g/mol
MDL Number: MFCD00671536
CAS Number: 73477-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR