D2R [(L-Asp)2 rhodamine 110] ≥98%, high purity
Synonyms: 2-[3-[[(2S)-2-[[(2S)-2-AMINO-4-HYDROXY-4-OXOBUTANOYL]AMINO]-4-HYDROXY-4-OXOBUTANOYL]AMINO]-6-IMINOXANTHEN-9-YL]BENZOIC ACID, 2,2,2-TRIFLUOROACETIC ACID(L-Asp)2 rhodamine 110.TFA salt
260-094-5001
89146-826EA 680.83 USD
89146-826
D2R [(L-Asp)2 rhodamine 110] ≥98%, high purity
D2R [(L-Asp)2 rhodamine 110]
Caspase substrate
Substrate for detection of intracellular caspase activation by flow cytometry and fluorescence microscopy after apoptosis induction in intact hematopoietic cell lines.