Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bel-Art Bulk-Packed Elliptical (Egg-Shaped) Spinbar Magnetic Stirring Bars, SP Scienceware

Supplier: Bel-Art Products
37130-9001 37130-9004 37130-9038 37130-9058 37130-9008 37130-9006 37130-9007
These egg-shaped stirring bars are designed especially for use in round-bottom vessels as they spin unrestricted by the vessel contour.

  • Particularly well suited for round bottom flasks; shape ensures complete mixing
  • Offers minimal contact when used in plastic containers
  • Egg shape provides pivot point without a separate ring or abrupt change in contour
  • Autoclave safe

Magnet itself is egg-shaped encapsulated in FDA grade, USP Class VI Teflon® PTFE, except Models 37130-9038, 37130-9004, and 37130-9058 which have a cylindrical magnet in egg-shaped Teflon® PTFE.

Certifications: Manufactured in registered ISO 9001:2008 facilities, as verified by SGS certification.



 

ORDER

Expand All / Collapse All
Fits Flask Size Diameter Length Supplier No. VWR Catalog Number Unit Price Quantity
10 and 25 mL (Round Bottom) 4.7 mm (3/16") 9.5 mm (3/8") 37130-9038 89089-574 Pack of 100 Retrieving
10 and 25 mL (Round Bottom) 6.3 mm (1/4") 15.9 mm (5/8") 37130-9058 10767-160 Case of 100 Retrieving
10, 25, 50, 100 mL (Round Bottom) 9.5 mm (3/8") 19 mm (3/4") 37130-9004 10767-156 Case of 100 Retrieving
100 and 200 mL (Round Bottom) 12.7 mm (1/2") 25.4 mm (1") 37130-9001 89089-564 Pack of 100 Retrieving
300 and 500 mL (Round Bottom) 15.9 mm (5/8") 31.8 mm (1 1/4") 37130-9006 89089-568 Pack of 100 Retrieving
500 and 1000 mL (Round Bottom) 19 mm (3/4") 41.3 mm (1 5/8") 37130-9007 89089-570 Pack of 100 Retrieving
3000 and 5000 mL (Round Bottom) 19 mm (3/4") 63.5 mm (2 1/2") 37130-9008 89089-572 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty