Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bel-Art Bulk-Packed Color Octagon Spinbar Magnetic Stirring Bars, SP Scienceware

Supplier: Bel-Art Products
37109-9015 37109-9014 37109-9018 37109-9017 37109-9012 37109-9011
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape

  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® brand PTFE Coating – Superior Chemical Compatibility
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable

Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars. TEFLON® is a registered trademark of The Chemours Company FC LLC and used under license by Bel-Art Products, Inc.

Certifications: Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification



 

ORDER

Expand All / Collapse All
Diameter Length Color Supplier No. VWR Catalog Number Unit Price Quantity
8 mm (5/16") 25.4 mm (1") Blue 37109-9012 89089-512 Pack of 100 Retrieving
8 mm (5/16") 25.4 mm (1") Yellow 37109-9011 89089-510 Pack of 100 Retrieving
8 mm (5/16") 38.1 mm (1 1/2") Blue 37109-9015 89089-516 Pack of 100 Retrieving
8 mm (5/16") 38.1 mm (1 1/2") Yellow 37109-9014 89089-514 Pack of 100 Retrieving
8 mm (5/16") 50.8 mm (2") Yellow 37109-9017 89089-518 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty