Laboratory Combination Showers, Justrite®
30107 30108
76183-652EA 2253.58 USD
76183-652 76183-650
Laboratory Combination Showers, Justrite®
Justrite Safety Showers provide the first line of defense against potential workplace hazards.
  • Powder coated stainless steel pipework, bowl, and rose ensures resistance to corrosive and flammable materials
  • Easy to clean
  • Pull handle shower activation, push plate eye/face wash activation
  • Chrome plated brass with stainless steel mesh eye/face wash diffusers
  • High-performance quarter-turn ball valves made of 316 L stainless steel endure the harshest environments and site conditions
  • Includes a universal eye wash and safety shower sign for enhanced visibility
  • Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements
Ideal for laboratories, schools, and general light-duty indoor use, these emergency combination showers are suitable for use in applications that are not subject to the possibility of freezing or overheating
The system features specially designed valves with low operating torque offering reliable, easy on/off operation when emergencies occur. Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure. Durable, coated stainless steel shower pull handle with nylon valve connection is easy to grip and activate for immediate water drenching from the shower rose that is designed for high flow rates.
Open bowl eye/face wash with high, ANSI-compliant flow rate includes aerated diffusers with fine mesh strainers and an individual flow controller which provide a soft, scrubbing action to the eyes and face. Features twin spray nozzles with dust covers that pop-off when eye wash is activated via the push plate. Easy-to-reach flow regulator valve adjusts to suit the severity of the accident. Highly visible red push plate with universal hand and eye wash sign for simple activation. Includes an inspection record tag for recording test and service history. Add optional foot treadle for a secondary means of eyewash activation. Some assembly may be required.
Showers are availablel in wall mount or floor mount designs.
Certifications: ANSI Z358.1-2014 and EN15154 compliant.
Delivery information: Ground shipment.