Handheld Eye/Face Wash, Wall Mount, with (2) 45° Nozzles, Justrite®
30016
76183-570EA 668.29 USD
76183-570
Handheld Eye/Face Wash, Wall Mount, with (2) 45° Nozzles, Justrite®
Justrite Safety Showers provide the first line of defense against potential workplace hazards.
  • Fitted with pop-off dust covers, a non-return valve, and flow regulator
  • Complete with 4.9 feet (1.5 m) of reinforced hose, particularly useful for treating disabled personnel
  • Squeeze grip activation operated with lock-on actuating button
Wall mounted hand-held eye/face wash features 45° angled double nozzles for hazardous environments where there is a risk of small spillages of chemicals or dangerous substances
The unit features a polyarylamide body (reinforced with glass fibers), powder coated brass nozzles, aned EPDM eye cups.
Meets ANSI and EN 15154 water flow standards for an eye/face wash. Includes an inspection record tag for recording test and service history.
Delivery information: Ground shipment.