Anti-TSPYL1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-86898
103269-664EA 767.69 USD
103269-664
Anti-TSPYL1 Rabbit Polyclonal Antibody
Antibodies
The TSPYL1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSPYL1. This antibody reacts with human. The TSPYL1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TSPYL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR