Anti-TSPAN8/TM4SF3 Mouse Monoclonal Antibody [clone: 1E5.]

Supplier: Novus Biologicals
H00007103-M02
103333-274EA 429 USD
103333-274
Anti-TSPAN8/TM4SF3 Mouse Monoclonal Antibody [clone: 1E5.]
Antibodies
The TSPAN8 / TM4SF3 Antibody (1E5.) from Novus Biologicals is a mouse monoclonal antibody to TSPAN8 / TM4SF3. This antibody reacts with human. The TSPAN8 / TM4SF3 Antibody (1E5.) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: TSPAN8/TM4SF3
Clonality: Monoclonal
Clone: 1E5.
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR