Anti-TSPAN3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-20604
102712-078EA 635.51 USD
102712-078
Anti-TSPAN3 Rabbit Polyclonal Antibody
Antibodies
The TSPAN3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSPAN3. This antibody reacts with human, mouse. The TSPAN3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TSPAN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR