Anti-TSPAN2 Mouse Monoclonal Antibody [clone: 1F2]

Supplier: Novus Biologicals
H00010100-M05
103338-138EA 798.46 USD
103338-138
Anti-TSPAN2 Mouse Monoclonal Antibody [clone: 1F2]
Antibodies
The TSPAN2 Antibody (1F2) from Novus Biologicals is a mouse monoclonal antibody to TSPAN2. This antibody reacts with human. The TSPAN2 Antibody (1F2) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: TSPAN2
Clonality: Monoclonal
Clone: 1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR